Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Bis[alpha-(4-chlorophenyl)benzyl]piperazine dihydrochloride, 97%

Catalog No. AAH6396106
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6396106 5 g
AAH6396103 1 g
AAH63961MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6396106 Supplier Thermo Scientific Chemicals Supplier No. H6396106
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 346451-15-8 | C30H30Cl4N2 | 560.384 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 346451-15-8
Molecular Formula C30H30Cl4N2
Molecular Weight (g/mol) 560.384
MDL Number MFCD03844641
InChI Key DLEQNCKNBPYHEY-UHFFFAOYSA-N
Synonym 1,4-bis 4-chlorophenyl phenylmethyl piperazine dihydrochloride, cetirizine impurity d, 1,4-bis 4-chlorophenyl phenylmethyl-piperazine dihydrochloride, 1,4-bis 4-chlorophenyl phenyl methyl piperazine dihydrochloride, 1,4-bis 4-chlorophenyl-phenylmethyl piperazine;dihydrochloride
PubChem CID 24720891
IUPAC Name 1,4-bis[(4-chlorophenyl)-phenylmethyl]piperazine;dihydrochloride
SMILES C1CN(CCN1C(C2=CC=CC=C2)C3=CC=C(C=C3)Cl)C(C4=CC=CC=C4)C5=CC=C(C=C5)Cl.Cl.Cl

Specifications

Color White
Quantity 5 g
Formula Weight 560.38
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1,4-Bis[alpha-(4-chlorophenyl)benzyl]piperazine dihydrochloride
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.