Learn More
Medchemexpress LLC 1,4-Bis(diphenylphosphino)butane | 7688-25-7 | 426.47 | 25 G
SDP
Catalog No. 5000448199
Encompass_Preferred
1,4-Bis(diphenylphosphino)butane is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
  • Used in Suzuki reaction
  • Essential ligand in palladium(0)-catalyzed acylcyanation of arylacetylenes
  • Plays a role in deprotection of allyloxycarbamates
  • Ligand for alkylations and isomerization reactions
  • Useful in organometallic chemistry
  • Applied in Heck reaction, Negishi coupling, and Sonogashira coupling
  • For research use only
Catalog No. 50-004-48199 Supplier Medchemexpress LLC Supplier No. 25GHYW007569
Add to Cart
Add to Cart
1,4-Bis(diphenylphosphino)butane is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
  • Used in Suzuki reaction
  • Essential ligand in palladium(0)-catalyzed acylcyanation of arylacetylenes
  • Plays a role in deprotection of allyloxycarbamates
  • Ligand for alkylations and isomerization reactions
  • Useful in organometallic chemistry
  • Applied in Heck reaction, Negishi coupling, and Sonogashira coupling
  • For research use only