Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Bis(hexafluoro-alpha-hydroxyisopropyl)benzene Hydrate 98.0+%, TCI America™
SDP

Catalog No. B14195G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B14195G 5 g
B141925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B14195G Supplier TCI America Supplier No. B14195G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1992-15-0
Molecular Formula C12H6F12O2
Molecular Weight (g/mol) 410.16
MDL Number MFCD00042091
InChI Key YTJDSANDEZLYOU-UHFFFAOYSA-N
Synonym 1,4-bis hexafluoro-alpha-hydroxyisopropyl benzene, 2,2'-1,4-phenylene bis 1,1,1,3,3,3-hexafluoropropan-2-ol, 1,4-bis 2-hydroxyhexafluoroisopropyl benzene, 1,4-bis perfluoro 2-hydroxyprop-2-yl benzene, 1,1,1,3,3,3-hexafluoro-2-4-1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl phenyl propan-2-ol, 1,4-bis 1,1,1,3,3,3-hexafluoro-2-hydroxypropyl benzene, alpha,alpha,alpha',alpha'-tetrakis trifluoromethyl-1,4-benzenedimethanol, 1,1,1,3,3,3-hexafluoro-2-4-2,2,2-trifluoro-1-hydroxy-1-trifluoromethyl ethy l phenyl propan-2-ol, 2,2'-p-phenylene bis 1,1,1,3,3,3-hexafluoro-2-propanol, 1,4-bis 2-hydroxyhexafluoro-2-propyl benzene
PubChem CID 631065
IUPAC Name 1,1,1,3,3,3-hexafluoro-2-[4-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)phenyl]propan-2-ol
SMILES OC(C1=CC=C(C=C1)C(O)(C(F)(F)F)C(F)(F)F)(C(F)(F)F)C(F)(F)F

Specifications

Melting Point 94°C
Color White
Quantity 5 g
Formula Weight 410.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Bis(hexafluoro-alpha-hydroxyisopropyl)benzene Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.