Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,4-Bis-maleimidobutane | 28537-70-4 | 98.0% | 248.23 | 250 MG
SDP

Catalog No. 5000451378
Encompass_Preferred

1,4-Bis-maleimidobutane is a biochemical reagent used for research. This compound appears as a white to off-white solid with a molecular weight of 248.23 and a purity of 98.0%. It has a CAS number of 28537-70-4.

  • Used as a biochemical reagent
  • White to off-white solid
  • Purity of 98.0%
  • Molecular weight of 248.23
  • CAS number: 28537-70-4

Catalog No. 50-004-51378 Supplier Medchemexpress LLC Supplier No. 250MGHYW011957
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,4-Bis-maleimidobutane is a biochemical reagent used for research. This compound appears as a white to off-white solid with a molecular weight of 248.23 and a purity of 98.0%. It has a CAS number of 28537-70-4.

  • Used as a biochemical reagent
  • White to off-white solid
  • Purity of 98.0%
  • Molecular weight of 248.23
  • CAS number: 28537-70-4

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.