Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Bis(methoxymethyl)benzene 99.0+%, TCI America™
SDP

Catalog No. B153025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B153025G 25 g
B1530500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B153025G Supplier TCI America Supplier No. B153025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6770-38-3
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD00025903
InChI Key DAJPMKAQEUGECW-UHFFFAOYSA-N
Synonym p-Xylene Glycol Dimethyl Ether
PubChem CID 81239
IUPAC Name 1,4-bis(methoxymethyl)benzene
SMILES COCC1=CC=C(C=C1)COC

Specifications

Color Colorless
Boiling Point 235°C
Quantity 25 g
Formula Weight 166.22
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,4-Bis(methoxymethyl)benzene
EINECSNumber (3)-2883
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.