Learn More
Medchemexpress LLC 1-(4-(bis(methyl-13C-d3)amino)phenyl)-2-bromoethan-1-one | 98.1% | 250.13 g/mol | C10H6D6BrNO (2x13C) | 50 MG
SDP
Catalog No. 5000282621
Encompass_Preferred
4-(Dimethylamino)phenacyl bromide-13C2,d6 is a stable-isotope labeled reagent used in analytical and synthetic research. As the 13C2, d6 isotopologue of 4-(dimethylamino)phenacyl bromide, it is intended for use as an internal standard, tracer, or reference material in mass spectrometry and method development. Provided as a solid with purity verified by HPLC.
  • Isotopically labeled with two 13C and six deuterium atoms
  • High purity: 98.1% by HPLC
  • Suitable as an internal standard for mass spectrometry
  • Useful for tracer and mechanistic studies
  • Available in small-mass vials for analytical applications
Catalog No. 50-002-82621 Supplier Medchemexpress LLC Supplier No. HYW128417S50MG
4-(Dimethylamino)phenacyl bromide-13C2,d6 is a stable-isotope labeled reagent used in analytical and synthetic research. As the 13C2, d6 isotopologue of 4-(dimethylamino)phenacyl bromide, it is intended for use as an internal standard, tracer, or reference material in mass spectrometry and method development. Provided as a solid with purity verified by HPLC.
  • Isotopically labeled with two 13C and six deuterium atoms
  • High purity: 98.1% by HPLC
  • Suitable as an internal standard for mass spectrometry
  • Useful for tracer and mechanistic studies
  • Available in small-mass vials for analytical applications