Learn More
Medchemexpress LLC 1-(4-(bis(methyl-13C-d3)amino)phenyl)-2-bromoethan-1-one | MFCD06409221 | 98.1% | 250.13 | C813C2H6D6BrNO | 10 MG
SDP
Catalog No. 5000282839
Encompass_Preferred
4-(Dimethylamino)phenacyl bromide-13C2,d6 is a 13C- and deuterium-labelled analogue of 4-(dimethylamino)phenacyl bromide supplied for laboratory research as an analytical tracer or internal standard. It is intended for research use only and not for human or clinical applications.
  • Isotopically labelled with 13C and deuterium for mass spectrometry tracing.
  • High purity (98.1%) suitable for analytical workflows.
  • Solid form with light yellow to yellow appearance for convenient handling.
  • Available in small milligram quantities for analytical and metabolism studies.
  • Stable when stored as powder under recommended temperatures.
Catalog No. 50-002-82839 Supplier Medchemexpress LLC Supplier No. HYW128417S10MG
4-(Dimethylamino)phenacyl bromide-13C2,d6 is a 13C- and deuterium-labelled analogue of 4-(dimethylamino)phenacyl bromide supplied for laboratory research as an analytical tracer or internal standard. It is intended for research use only and not for human or clinical applications.
  • Isotopically labelled with 13C and deuterium for mass spectrometry tracing.
  • High purity (98.1%) suitable for analytical workflows.
  • Solid form with light yellow to yellow appearance for convenient handling.
  • Available in small milligram quantities for analytical and metabolism studies.
  • Stable when stored as powder under recommended temperatures.