Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,4-Bis(Triethoxysilyl)Benzene

Catalog No. 5980385G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
20 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
59-803-85G 5 g
59-803-820G 20 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 59-803-85G Supplier Sigma Aldrich Supplier No. 5980385G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,4-Bis(triethoxysilyl)benzene (BTEB) is a silicon-based nucleophile shown to be reactive in Pd-catalyzed cross-coupling reactions. It is extensively used as a precursor in the synthesis of a variety of periodic mesoporous organosilicas (PMOs).

Specifications

CAS 2615-18-1
Density 1.015 g/mL at 25°C
Boiling Point 130°C to 133°C (0.5 mmHg, lit.)
Molecular Formula C18H34O6Si2
Refractive Index n20/D 1.456
Linear Formula C6H4(Si(OC2H5)3)2
MDL Number MFCD05664351
Quantity 5 g
Synonym BTEB
Molecular Weight (g/mol) 402.63 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.