Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Bisbenzil 97.0+%, TCI America™
SDP

Catalog No. B14805G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B14805G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B14805G Supplier TCI America Supplier No. B14805G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3363-97-1
Molecular Formula C22H14O4
Molecular Weight (g/mol) 342.35
MDL Number MFCD00039561
InChI Key FUEGWHHUYNHBNI-UHFFFAOYSA-N
Synonym 1,4-Bis(benzoylcarbonyl)benzene
PubChem CID 137905
IUPAC Name 1-[4-(2-oxo-2-phenylacetyl)phenyl]-2-phenylethane-1,2-dione
SMILES O=C(C(=O)C1=CC=C(C=C1)C(=O)C(=O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 125°C
Color Yellow
Quantity 5 g
Formula Weight 342.35
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Bisbenzil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.