Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Bromophenyl)-1,2,2-triphenylethylene 98.0+%, TCI America™
SDP

Catalog No. B36341G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36341G 1 g
B36345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36341G Supplier TCI America Supplier No. B36341G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 34699-28-0
Molecular Formula C26H19Br
Molecular Weight (g/mol) 411.34
MDL Number MFCD20257904
InChI Key MYJLJYSALGARCM-UHFFFAOYSA-N
Synonym 1-Bromo-4-(1,2,2-triphenylethenyl)benzene
PubChem CID 24882208
IUPAC Name 1-bromo-4-(1,2,2-triphenylethenyl)benzene
SMILES BrC1=CC=C(C=C1)C(=C(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 159°C
Color White-Yellow
Quantity 1 g
Formula Weight 411.34
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(4-Bromophenyl)-1,2,2-triphenylethylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.