Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Bromophenyl)-1,3-butanedione 98.0+%, TCI America™
SDP

Catalog No. B47541G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47541G 1 g
B47545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B47541G Supplier TCI America Supplier No. B47541G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4023-81-8
Molecular Formula C10H9BrO2
Molecular Weight (g/mol) 241.08
MDL Number MFCD04619579
InChI Key HPMLNAUGJOJXLT-POHAHGRESA-N
Synonym (4-Bromobenzoyl)acetone
PubChem CID 432653
IUPAC Name (3Z)-4-(4-bromophenyl)-4-hydroxybut-3-en-2-one
SMILES CC(=O)\C=C(/O)C1=CC=C(Br)C=C1

Specifications

Melting Point 93°C
Color White-Yellow
Quantity 1 g
Formula Weight 241.08
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(4-Bromophenyl)-1,3-butanedione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.