Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Bromophenyl)naphthalene 98.0+%, TCI America™
SDP

Catalog No. B39781G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39781G 1 g
B39785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B39781G Supplier TCI America Supplier No. B39781G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 204530-94-9
Molecular Formula C16H11Br
Molecular Weight (g/mol) 283.168
MDL Number MFCD07369916
InChI Key YRPIGRRBBMFFBE-UHFFFAOYSA-N
Synonym 1-4-bromophenyl naphthalene, 1-4-bromo-phenyl-naphthalene, 1-4-bromophenyl-naphthlene, 1-4-bromophenyl-naphthalene, naphthalene, 1-4-bromophenyl, 1-4-bromophenyl naphthlene, pubchem13401, 1 4-bromophenyl naphthalene, 4-1-naphthyl bromobenzene
PubChem CID 11208338
IUPAC Name 1-(4-bromophenyl)naphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2C3=CC=C(C=C3)Br

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 283.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(4-Bromophenyl)naphthalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.