Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Bromophenyl)pyrrolidine 98.0+%, TCI America™
SDP

Catalog No. B39005G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39005G 5 g
B39001G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B39005G Supplier TCI America Supplier No. B39005G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 22090-26-2
Molecular Formula C10H12BrN
Molecular Weight (g/mol) 226.117
MDL Number MFCD04112480
InChI Key BVEGBJXZICCEQW-UHFFFAOYSA-N
Synonym 1-4-bromophenyl pyrrolidine, n-4-bromophenyl pyrrolidine, 1-4-bromophenyl-pyrrolidine, acmc-209frt, 4-pyrrolidinobromobenzene, 4-bromophenyl pyrrolidine, 1-4-bromo-phenyl-pyrrolidine, pyrrolidine,1-4-bromophenyl, 4-bromo-1-1-pyrrolidinyl benzene
PubChem CID 7016457
IUPAC Name 1-(4-bromophenyl)pyrrolidine
SMILES C1CCN(C1)C2=CC=C(C=C2)Br

Specifications

Melting Point 107°C
Color White
Quantity 5 g
Formula Weight 226.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(4-Bromophenyl)pyrrolidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.