Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, 1-(4-Bromopyridin-2-yl)ethanone, 98%, [1060805-69-7], Y14744 200.033 C7H6BrNO

Catalog No. 501456308
Encompass_Preferred

Compound: 1-(4-Bromopyridin-2-yl)ethanone

CAS#: 1060805-69-7

MDL#: MFCD13189212

Background: A brominated pyridine bearing a reactive acetyl (methyl ketone) group, used as a building block in pharmaceutical research. The bromine offers a reactive site for cross-coupling, while the ketone allows for reductive amination and other carbonyl chemistry.

Storage: Store at 2-8°C, under dry conditions.

Catalog No. 50-145-6308 Supplier Frontier Specialty Chemicals Supplier No. Y147445G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 1-(4-Bromopyridin-2-yl)ethanone

CAS#: 1060805-69-7

MDL#: MFCD13189212

Background: A brominated pyridine bearing a reactive acetyl (methyl ketone) group, used as a building block in pharmaceutical research. The bromine offers a reactive site for cross-coupling, while the ketone allows for reductive amination and other carbonyl chemistry.

Storage: Store at 2-8°C, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.