Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,4-Butanediol diacrylate

Catalog No. 41174425ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
100 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-174-425ML 25 mL
41-174-4100ML 100 mL
41-174-41L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 41-174-425ML Supplier Sigma Aldrich Supplier No. 41174425ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,4-Butanediol diacrylate may be used as a multifunctional crosslinker.

Specifications

CAS 1070-70-8
Density 1.051 g/mL (at 25°C)
Boiling Point 83°C (0.3 mmHg) (literature)
Molecular Formula C10H14O4
Refractive Index n20/D 1.456 (literature)
Linear Formula (H2C=CHCO2CH2CH2-)2
Quantity 25 mL
MDL Number MFCD00014940
Synonym 1,4-Bis(acryloyloxy)butane; Tetramethylene diacrylate
Molecular Weight (g/mol) 198.22
Percent Purity 87%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.