Learn More
Chemglass Life Sciences Cajon Unions Only, Use with 1/4" OD Tube, Airfree, Schlenk
SDP
Catalog No. AF006704
Encompass_Preferred
Cajon Unions Only, Use with 1/4" OD Tube, Airfree, Schlenk. Unions provide seals directly from the listed sizes on a glass to glass basis, or can be used with the AF-0068 cuffs and flexible tubing. They can be used with glass tubing as much as 1/32in undersize. Unions are made from #316 stainless steel. O-Rings are 70 durometer Viton.
Catalog No. AF006704 Supplier Chemglass Life Sciences Supplier No. AF006704
Cajon Unions Only, Use with 1/4" OD Tube, Airfree, Schlenk. Unions provide seals directly from the listed sizes on a glass to glass basis, or can be used with the AF-0068 cuffs and flexible tubing. They can be used with glass tubing as much as 1/32in undersize. Unions are made from #316 stainless steel. O-Rings are 70 durometer Viton.