Learn More
AdipoGen 1-4-Carboxybutyl-3 7-dmx
SDP
Supplier:  AdipoGen CDXC0312M025
Encompass_Preferred
Chemical. CAS 38975-44-9. Formula C12H16N4O4. MW 280.28. Synthetic Pentoxifylline PTX metabolite. Can be used as reference material. Showed platelet aggregation in whole blood in vitro. Shown to be more effective than PTX in protecting WC/1 murine fibrosarcoma cells from TNF-alpha cytotoxicity and in inhibiting neutrophil superoxide anion production, degranulation lactoferrin release and surface expression of the beta-2 integrin CD11b/CD18.
Catalog No. 50-256-1384