Learn More
Medchemexpress LLC 1-[4-chloro-3-(trifluoromethyl)phenyl]-3-(2,2-difluoro-2H-1,3-benzodioxol-5-yl)urea | 2165324-62-7 | MFCD32667017 | 99.4% | 394.68 g·mol⁻¹ | C15H8ClF5N2O3 | 5 MG
SDP
Catalog No. 5000262170
Encompass_Preferred
A small-molecule research compound used in antibacterial and pharmacology studies. It is described as a sorafenib analogue with reported oral bioavailability and activity against pathogenic Staphylococcus aureus strains. Supplied as a solid for laboratory research.
  • Small-molecule sorafenib analogue suitable for in vitro and in vivo studies.
  • Reported oral bioavailability and antibacterial activity against Staphylococcus aureus strains.
  • High purity (≈99.4%) for consistent experimental results.
  • Molecular weight 394.68 g·mol⁻¹ and formula C15H8ClF5N2O3 for reference.
  • Supplied as a white to off-white solid with recommended cold storage.
  • Available in small research pack sizes suitable for experimental use.
Catalog No. 50-002-62170 Supplier Medchemexpress LLC Supplier No. HY1331195MG
A small-molecule research compound used in antibacterial and pharmacology studies. It is described as a sorafenib analogue with reported oral bioavailability and activity against pathogenic Staphylococcus aureus strains. Supplied as a solid for laboratory research.
  • Small-molecule sorafenib analogue suitable for in vitro and in vivo studies.
  • Reported oral bioavailability and antibacterial activity against Staphylococcus aureus strains.
  • High purity (≈99.4%) for consistent experimental results.
  • Molecular weight 394.68 g·mol⁻¹ and formula C15H8ClF5N2O3 for reference.
  • Supplied as a white to off-white solid with recommended cold storage.
  • Available in small research pack sizes suitable for experimental use.