Learn More
Medchemexpress LLC 1-[4-chloro-3-(trifluoromethyl)phenyl]-3-(2,2-difluoro-2H-1,3-benzodioxol-5-yl)urea | 2165324-62-7 | MFCD32667017 | 99.4% | C15H8ClF5N2O3 | 10MG
SDP
Catalog No. 5000259196
Encompass_Preferred
PK150 is an orally bioavailable Sorafenib analogue that exhibits potent antibacterial activity against Staphylococcus aureus strains, including MSSA, MRSA, and VISA, with minimum inhibitory concentrations in the submicromolar range. It is supplied for research use and is intended for biochemical and microbiological studies.
  • Orally bioavailable Sorafenib analogue.
  • Active against MSSA, MRSA, and VISA at submicromolar MICs.
  • High reported purity suitable for research applications.
  • Available in multiple small-scale packaging sizes for laboratory use.
  • Chemical formula C15H8ClF5N2O3, molecular weight 394.68.
Catalog No. 50-002-59196 Supplier Medchemexpress LLC Supplier No. HY13311910MG
PK150 is an orally bioavailable Sorafenib analogue that exhibits potent antibacterial activity against Staphylococcus aureus strains, including MSSA, MRSA, and VISA, with minimum inhibitory concentrations in the submicromolar range. It is supplied for research use and is intended for biochemical and microbiological studies.
  • Orally bioavailable Sorafenib analogue.
  • Active against MSSA, MRSA, and VISA at submicromolar MICs.
  • High reported purity suitable for research applications.
  • Available in multiple small-scale packaging sizes for laboratory use.
  • Chemical formula C15H8ClF5N2O3, molecular weight 394.68.