Learn More
Medchemexpress LLC 1-(4-Chlorobenzhydryl)piperazine | 303-26-4 | 286.80 | 1 ML
SDP
Catalog No. 5000447089
Encompass_Preferred
1-(4-Chlorobenzhydryl)piperazine is a high-purity biochemical reagent, offered as a 10 mM solution in DMSO, suitable for life science research. This compound is known as an inactive metabolite of meclizine and chlorcyclizine, and can be found as an impurity in commercial preparations of hydroxyzine and cetirizine. It is stable under recommended storage conditions and appears as a white to off-white solid.
  • Used as a biological material or organic compound for life science research
  • Inactive metabolite of meclizine and chlorcyclizine
  • Found as an impurity in commercial preparations of hydroxyzine and cetirizine
  • Utilized in the synthesis of voltage-gated sodium channel 1.7 inhibitors
  • Applied in the synthesis of anticancer agents
  • Employed in the synthesis of antihistamines
Catalog No. 50-004-47089 Supplier Medchemexpress LLC Supplier No. HYW00197810MM/1ML
Add to Cart
Add to Cart
1-(4-Chlorobenzhydryl)piperazine is a high-purity biochemical reagent, offered as a 10 mM solution in DMSO, suitable for life science research. This compound is known as an inactive metabolite of meclizine and chlorcyclizine, and can be found as an impurity in commercial preparations of hydroxyzine and cetirizine. It is stable under recommended storage conditions and appears as a white to off-white solid.
  • Used as a biological material or organic compound for life science research
  • Inactive metabolite of meclizine and chlorcyclizine
  • Found as an impurity in commercial preparations of hydroxyzine and cetirizine
  • Utilized in the synthesis of voltage-gated sodium channel 1.7 inhibitors
  • Applied in the synthesis of anticancer agents
  • Employed in the synthesis of antihistamines