Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Chlorophenyl)-1-cyclopentanecarbonyl chloride, 97%, Thermo Scientific™

Catalog No. AC135020250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC135020250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC135020250 Supplier Thermo Scientific Chemicals Supplier No. 135020250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71501-44-5
Molecular Formula C12H12Cl2O
Molecular Weight (g/mol) 243.127
InChI Key RZHJJILZUKNEKM-UHFFFAOYSA-N
Synonym 1-4-chlorophenyl cyclopentanecarbonyl chloride, 1-4-chlorophenyl-1-cyclopentanecarbonyl chloride, 1-4-chlorophenyl-1-cyclopentanecarbonylchloride, 1-4-chlorophenyl cyclopentane-1-carbonyl chloride, cyclopentanecarbonylchloride, 1-4-chlorophenyl, 1-4-chlorophenyi-1-cyclopentanecarbonyl chloride, cyclopentanecarbonyl chloride, 1-4-chlorophenyl, 1-4-chlorophenyl-1-cyclopentanecarboxylic acid chloride, 1-4-chlorophenyl cyclopentane-1-carboxylic acid chloride, cyclopentanecarbonyl chloride,1-4-chlorophenyl
PubChem CID 2723774
IUPAC Name 1-(4-chlorophenyl)cyclopentane-1-carbonyl chloride
SMILES C1CCC(C1)(C2=CC=C(C=C2)Cl)C(=O)Cl

Specifications

Quantity 25 g
Refractive Index 1.5545 to 1.5565
Formula Weight 243.13
Chemical Name or Material 1-(4-Chlorophenyl)-1-cyclopentanecarbonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.