Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Chlorophenyl)-1-cyclopentanecarboxylic acid, 98%, Thermo Scientific™

Catalog No. AC161571000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC161571000 100 g Glass bottle
AC161570050 5 g Glass bottle
AC161570250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC161571000 Supplier Thermo Scientific Chemicals Supplier No. 161571000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 80789-69-1
Molecular Formula C12H13ClO2
Molecular Weight (g/mol) 224.69
MDL Number MFCD00001373
InChI Key QJNFJEMGWIQMJT-UHFFFAOYSA-N
Synonym 1-4-chlorophenyl cyclopentanecarboxylic acid, 1-4-chlorophenyl-1-cyclopentanecarboxylic acid, 1-4-chlorophenyl cyclopentane-1-carboxylic acid, cyclopentanecarboxylic acid, 1-4-chlorophenyl, asischem d50961, timtec-bb sbb000649, acmc-209pk2, labotest-bb lt00453384, 1-4-chloro phenyl cyclopentanecarboxylic acid, 1-4-chloro-phenyl-cyclopentylcarboxylic acid
PubChem CID 97447
IUPAC Name 1-(4-chlorophenyl)cyclopentane-1-carboxylic acid
SMILES C1CCC(C1)(C2=CC=C(C=C2)Cl)C(=O)O

Specifications

Melting Point 160.0°C to 164.0°C
Color Cream to Gray
CAS Min % 97.5
CAS Max % 100.0
Quantity 100 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Powder or Crystals
Assay Percent Range 98%
Linear Formula ClC6H4C5H8CO2H
Formula Weight 224.69
Percent Purity 98%
Chemical Name or Material 1-(4-Chlorophenyl)-1-cyclopentanecarboxylic acid
Show More Show Less
EINECSNumber 279-552-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.