Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Chlorophenyl)-2-(isopropylamino)ethanol, 98%, Thermo Scientific™

Catalog No. AAH50756MD
Catalog No. AAH50756MD Supplier Thermo Scientific Chemicals Supplier No. H50756MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 23299-13-0
Molecular Formula C11H16ClNO
MDL Number MFCD08060970
Synonym 1-4-chlorophenyl-2-isopropylamino ethanol, 1-4-chloro-phenyl-2-isopropylamino-ethanol, acmc-20apb2, 1-4-chlorophenyl-2-isopropylaminoethanol, 1-4-chlorophenyl-2-methylethyl amino ethan-1-ol, 1-4-chlorophenyl-2-propan-2-yl amino ethan-1-ol, benzenemethanol,4-chloro-a-1-methylethyl amino methyl
InChI Key ODPACOSFGOJIQY-UHFFFAOYSA-N
SMILES CC(C)NCC(C1=CC=C(C=C1)Cl)O
IUPAC Name 1-(4-chlorophenyl)-2-(propan-2-ylamino)ethanol
Molecular Weight (g/mol) 213.705
PubChem CID 24212023
Formula Weight 213.71
Percent Purity 98%
Chemical Name or Material 1-(4-Chlorophenyl)-2-(isopropylamino)ethanol
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.