Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Chlorophenyl)-3-methyl-5-pyrazolone 98.0+%, TCI America™
SDP

Catalog No. C095325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C095325G 25 g
C0953500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C095325G Supplier TCI America Supplier No. C095325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13024-90-3
Molecular Formula C10H9ClN2O
Molecular Weight (g/mol) 208.65
MDL Number MFCD00043814
InChI Key WHIXQFSPEDIMGL-UHFFFAOYSA-N
PubChem CID 83055
IUPAC Name 1-(4-chlorophenyl)-3-methyl-4,5-dihydro-1H-pyrazol-5-one
SMILES CC1=NN(C(=O)C1)C1=CC=C(Cl)C=C1

Specifications

Melting Point 167°C
Quantity 25 g
Physical Form Crystal-Powder at 20°C
Formula Weight 208.65
Percent Purity ≥98.0% (T)
Chemical Name or Material 1-(4-Chlorophenyl)-3-methyl-5-pyrazolone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.