Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Chlorophenyl)cyclohexane-1-carboxylic acid, 95%

Catalog No. AAL0533606
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0533606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0533606 Supplier Thermo Scientific Chemicals Supplier No. L0533606

missing translation for 'validMainframeMsg'

CAS: 58880-37-8 | C13H14ClO2 | 237.70 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58880-37-8
Molecular Formula C13H14ClO2
Molecular Weight (g/mol) 237.70
MDL Number MFCD00019350
InChI Key UPNXUJXIIZGXLQ-UHFFFAOYSA-M
Synonym 1-4-chlorophenyl cyclohexanecarboxylic acid, 1-4-chlorophenyl cyclohexane-1-carboxylic acid, 1-4-chlorophenyl-1-cyclohexanecarboxylic acid, 1-4-chlorophenyl-1-cyclohexane-carboxylic acid, cyclohexanecarboxylic acid, 1-4-chlorophenyl, acmc-1cuiq, 4-chlorophenylcyclohexanecarboxylic acid, 4-chlorophenyl cyclohexanecarboxylic acid, 1-4-chlorophenyl cyclohexanecarboxylicacid
PubChem CID 100873
IUPAC Name 1-(4-chlorophenyl)cyclohexane-1-carboxylic acid
SMILES [O-]C(=O)C1(CCCCC1)C1=CC=C(Cl)C=C1

Specifications

Melting Point 151°C to 153°C
Quantity 5 g
Beilstein 3297868
Formula Weight 238.71
Percent Purity 95%
Chemical Name or Material 1-(4-Chlorophenyl)cyclohexane-1-carboxylic acid
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 261-481-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.