Learn More
Medchemexpress LLC 1-(4-Chlorophenyl)piperazine | 38212-33-8 | 196.68 | 100 G
SDP
Catalog No. 5000448988
Encompass_Preferred
1-(4-Chlorophenyl)piperazine is a chemical intermediate that can be used for the synthesis of D2R ligand. It is for research use only.
  • Chemical intermediate
  • Can be used for synthesis of D2R ligand
  • For research use only
  • Solid appearance
  • White to light yellow color
Catalog No. 50-004-48988 Supplier Medchemexpress LLC Supplier No. HYW004758100G
1-(4-Chlorophenyl)piperazine is a chemical intermediate that can be used for the synthesis of D2R ligand. It is for research use only.
  • Chemical intermediate
  • Can be used for synthesis of D2R ligand
  • For research use only
  • Solid appearance
  • White to light yellow color