Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-(4-Chlorophenyl)piperazine | 38212-33-8 | 196.68 | 25 G
SDP

Catalog No. 5000449857
Encompass_Preferred

1-(4-Chlorophenyl)piperazine is a chemical intermediate that can be used for the synthesis of D2R ligand.

  • Appearance: Solid
  • Color: White to light yellow
  • Solubility (In Vitro): DMSO: 100 mg/mL

Catalog No. 50-004-49857 Supplier Medchemexpress LLC Supplier No. 25GHYW004758
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-(4-Chlorophenyl)piperazine is a chemical intermediate that can be used for the synthesis of D2R ligand.

  • Appearance: Solid
  • Color: White to light yellow
  • Solubility (In Vitro): DMSO: 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.