Learn More
Medchemexpress LLC 1-(4-Chlorophenyl)piperazine | 38212-33-8 | 196.68 | 50 G
SDP
Catalog No. 5000445558
Encompass_Preferred
1-(4-Chlorophenyl)piperazine is a chemical intermediate used for the synthesis of D2R ligand. It is intended for research purposes only.
  • Solid appearance
  • White to light yellow color
  • Soluble in DMSO (100 mg/mL)
Catalog No. 50-004-45558 Supplier Medchemexpress LLC Supplier No. HYW00475850G
1-(4-Chlorophenyl)piperazine is a chemical intermediate used for the synthesis of D2R ligand. It is intended for research purposes only.
  • Solid appearance
  • White to light yellow color
  • Soluble in DMSO (100 mg/mL)