Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Chlorophenyl)piperazine dihydrochloride, 95%, Thermo Scientific™

Catalog No. AC109921000
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC109921000 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC109921000 Supplier Thermo Scientific Chemicals Supplier No. 109921000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38869-46-4
Molecular Formula C10H14Cl2N2
Molecular Weight (g/mol) 233.14
MDL Number MFCD00012766
InChI Key ZHGRQBSZTVJDHU-UHFFFAOYSA-N
Synonym 1-4-chlorophenyl piperazine dihydrochloride, 1-4-chlorophenyl-piperazine dihydrochloride, 1-4-chlorophenyl piperazine 2hcl, para-chlorophenylpiperazine dihydrochloride, piperazine, 1-4-chlorophenyl-, dihydrochloride, pubchem8586, acmc-1ctdy, ksc493m4r, 1-p-chlorophenyl piperazine, 4-chlorophenylpiperazine dihydrochloride
PubChem CID 12212341
IUPAC Name hydrogen 1-(4-chlorophenyl)piperazine chloride
SMILES [H+].[Cl-].ClC1=CC=C(C=C1)N1CCNCC1

Specifications

Melting Point 274.0°C to 279.0°C
Quantity 100 mL
Formula Weight 269.59
Chemical Name or Material 1-(4-Chlorophenyl)piperazine dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.