Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Chlorophenyl)piperazine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. C111725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C111725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C111725G Supplier TCI America Supplier No. C111725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13078-12-1
Molecular Formula C10H14Cl2N2
Molecular Weight (g/mol) 233.136
MDL Number MFCD00060187
InChI Key ZHGRQBSZTVJDHU-UHFFFAOYSA-N
Synonym 1-4-chlorophenyl piperazine hydrochloride, para-chlorophenylpiperazine hydrochloride, timtec-bb sbb003262, piperazine, 1-4-chlorophenyl-, monohydrochloride, 4-chlorophenyl piperazine, chloride, 1-4-chlorophenyl piperazine;hydrochloride, 1-4-chlorophenyl-piperazine hydrochloride, acmc-1c3h8, 1-4-chlorophenyl piperazine hcl, 1-4-chlorophenyl piperazinehydrochloride
PubChem CID 3084720
IUPAC Name 1-(4-chlorophenyl)piperazine;hydrochloride
SMILES C1CN(CCN1)C2=CC=C(C=C2)Cl.Cl

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 233.14
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1-(4-Chlorophenyl)piperazine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.