Learn More
Selleck Chemical LLC 1.4-Cineole S5419-25mg
SDP
Supplier:  Selleck Chemical LLC S541925MG
Encompass_Preferred
1 4-Cineole (Isocineole) is a widely distributed natural oxygenated monoterpene 1 4-Cineole present in eucalyptus oil activates both human TRPM8 and human TRPA1
Catalog No. 50-310-8800