Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,4-Cyclohexadiene

Catalog No. 289105ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-910-5ML 5 mL
28-910-25ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 28-910-5ML Supplier Sigma Aldrich Supplier No. 289105ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1,4-cyclohexadiene is an effective hydrogen donor for catalytic hydrogenation reaction
  • The interaction between graphene segments and 1,4-cyclohexadiene was stuided using density-functional tight-binding (dftb) method.1,4-cyclohexadiene (1,4-chd) was used to study the formation of parent ion from heavy fragmentation of 1,4-chd on irradiation with a high-intensity laser pulse.reagent for selectively cleaving benzyl esters in the presence of benzyl ethers by catalytic h-transfer

Specifications

CAS 628-41-1
Density 0.847 g/mL at 25°C (lit.)
Boiling Point 88°C to 89°C (lit.)
Molecular Formula C6H8
Linear Formula C6H8
Refractive Index n20/D 1.472 (lit.); n20/D 1.473
Quantity 5 mL
MDL Number MFCD00001535
Synonym 1,4-Dihydrobenzene
Molecular Weight (g/mol) 80.13
Percent Purity ≥97.0% (GC)
Biological Activity Blood
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.