Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,4-Cyclohexanedimethanol

Catalog No. 125598100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
3 kg
17 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-559-8100G 100 g
12-559-817KG 17 kg
12-559-83KG 3 kg
12-559-8500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 12-559-8100G Supplier Sigma Aldrich Supplier No. 125598100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1,4-cyclohexanedimethano is extensively used as cross-linking reagent in polymer industry.1,4-cyclohexanedimethanol has been used in the synthesis of polyketal copolymers
  • It was used as diol comonomer during the synthesis of polyester-carbonates based on 1,3-propylene-co-1,4-cyclohexanedimethylene succinate

Specifications

CAS 105-08-8
Boiling Point 283°C (lit.)
Molecular Formula C8H16O2
Linear Formula C6H10(CH2OH)2
MDL Number MFCD00001512
Quantity 100 g
Synonym 1,4-Bis(hydroxymethyl)cyclohexane, mixture of cis and trans
Molecular Weight (g/mol) 144.21
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.