Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Cyclohexanedimethanol Bis(2-ethylhexanoate) (cis- and trans- mixture) 98.0+%, TCI America™
SDP

Catalog No. C113325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C113325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C113325G Supplier TCI America Supplier No. C113325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 53148-32-6
Molecular Formula C24H44O4
Molecular Weight (g/mol) 396.612
MDL Number MFCD00060063
InChI Key WZCWEBVKSPNNNN-UHFFFAOYSA-N
PubChem CID 103744
IUPAC Name [4-(2-ethylhexanoyloxymethyl)cyclohexyl]methyl 2-ethylhexanoate
SMILES CCCCC(CC)C(=O)OCC1CCC(CC1)COC(=O)C(CC)CCCC

Specifications

Color Yellow
Quantity 25 g
Formula Weight 396.61
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,4-Cyclohexanedimethanol Bis(2-ethylhexanoate) (cis- and trans- mixture)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.