Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Cyclohexanedimethanol Diisovalerate (cis- and trans- mixture) 98.0+%, TCI America™
SDP

Catalog No. C11351G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C11351G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C11351G Supplier TCI America Supplier No. C11351G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 377085-57-9
Molecular Formula C18H32O4
MDL Number MFCD00060061

Specifications

Color Yellow
Quantity 1 g
Formula Weight 312.45
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,4-Cyclohexanedimethanol Diisovalerate (cis- and trans- mixture)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.