Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,4-Cyclohexanedimethanol divinyl ether, mixture of isomers

Catalog No. 406171100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-617-1100ML 100 mL
40-617-1500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 40-617-1100ML Supplier Sigma Aldrich Supplier No. 406171100ML
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,4-Cyclohexanedimethanol divinyl ether, mixture of isomers

Specifications

CAS 17351-75-6
Density 0.919 g/mL (at 25°C)
Boiling Point 126°C (4 mmHg)
Molecular Formula C12H20O2
Refractive Index n20/D 1.472 (literature)
Linear Formula C6H10(CH2OCH=CH2)2
Quantity 100 mL
MDL Number MFCD00080497
Synonym 1,4-Bis[(vinyloxy)methyl]cyclohexane
Molecular Weight (g/mol) 196.29
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.