Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Cyclohexanediol, cis + trans, 98%

Catalog No. AAB2044622
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2044622 100 g
AAB2044614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2044622 Supplier Thermo Scientific Chemicals Supplier No. B2044622
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 556-48-9 | C6H12O2 | 116.16 g/mol

1,4-Cyclohexanediol, cis + trans, is used as a chemical, organic, pharmaceutical intermediate. It is also used as an important intermediate for manufacturing of anti-Cancer and anti-Infective Drugs - dihydroartemisinin and dihydroartemisitene dimers. It is also used in co polyester resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Cyclohexanediol, cis + trans, is used as a chemical, organic, pharmaceutical intermediate. It is also used as an important intermediate for manufacturing of anti-Cancer and anti-Infective Drugs - dihydroartemisinin and dihydroartemisitene dimers. It is also used in co polyester resins.

Solubility
It is highly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, acid chlorides, acid anhydrides, strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 556-48-9
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00001448,MFCD00063612,MFCD00075462
InChI Key VKONPUDBRVKQLM-UHFFFAOYSA-N
Synonym 1,4-cyclohexanediol, cis-1,4-cyclohexanediol, trans-1,4-cyclohexanediol, trans-cyclohexane-1,4-diol, quinitol, 1,4-cyclohexanediol, trans, cis-cyclohexane-1,4-diol, 1,4-cyclohexanediol, cis, trans-1,4-dihydroxycyclohexane, cis-1,4-dihydroxycyclohexane
PubChem CID 11162
IUPAC Name cyclohexane-1,4-diol
SMILES OC1CCC(O)CC1

Specifications

Melting Point 98°C to 103°C
Boiling Point 149°C to 151°C (20 mmHg)
Flash Point 65°C (149°F)
Quantity 100 g
Beilstein 2036658
Solubility Information It is highly soluble in water.
Formula Weight 116.16
Percent Purity 98%
Chemical Name or Material 1,4-Cyclohexanediol, cis + trans
EINECSNumber 209-126-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.