Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Cyclohexanedione, 98%

Catalog No. p-7081218
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1110014 25 g
AAA1110022 100 g
AAA1110036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1110014 Supplier Thermo Scientific Chemicals Supplier No. A1110014
Only null left
Add to Cart

CAS: 637-88-7 | C6H8O2 | 112.128 g/mol

1,4-Cyclohexanedione is used in the preparation of 1,4 benzoquinone and bromoorganics. It is also used to study the influence of visible light on the bromate-1,4-cyclohexanedione-ferroin oscillating reaction. It plays a vital role in pharmaceuticals, plant growth regulator and as a conducting material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Cyclohexanedione is used in the preparation of 1,4 benzoquinone and bromoorganics. It is also used to study the influence of visible light on the bromate-1,4-cyclohexanedione-ferroin oscillating reaction. It plays a vital role in pharmaceuticals, plant growth regulator and as a conducting material.

Solubility
Soluble in methanol and ethanol.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 637-88-7
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.128
MDL Number MFCD00001606
InChI Key DCZFGQYXRKMVFG-UHFFFAOYSA-N
Synonym 1,4-cyclohexanedione, tetrahydroquinone, 1,4-dioxocyclohexane, 1,4-cyclohexandione, 1,4 cyclohexane dione, 1,4-cyclohexane-dione, cyclohexan-1,4-dione, pubchem13686, cyclohexane-1,4-quinone, acmc-1b8ar
PubChem CID 12511
ChEBI CHEBI:28286
IUPAC Name cyclohexane-1,4-dione
SMILES C1CC(=O)CCC1=O

Specifications

Melting Point 77°C to 78°C
Boiling Point 132°C (20 mmHg)
Flash Point 132°C (269°F)
Quantity 25 g
Beilstein 774152
Solubility Information Soluble in methanol and ethanol.
Formula Weight 112.13
Percent Purity 98%
Chemical Name or Material 1,4-Cyclohexanedione
EINECSNumber 211-306-0
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.