Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Cyclohexanedione Mono-2,2-dimethyltrimethylene Ketal 98.0+%, TCI America™
SDP

Catalog No. C232825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C232825G 25 g
C23285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C232825G Supplier TCI America Supplier No. C232825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 69225-59-8
Molecular Formula C11H18O3
Molecular Weight (g/mol) 198.262
MDL Number MFCD00006652
InChI Key COKVDTKAWIFNTH-UHFFFAOYSA-N
Synonym 3,3-dimethyl-1,5-dioxaspiro 5.5 undecan-9-one, 1,4-cyclohexanedione mono-2,2-dimethyltrimethylene ketal, 1,4-cyclohexanedione mono 2,2-dimethyltrimethylene ketal, 3,3-dimethyl-1,5-dioxa-spiro 5.5 undecan-9-one, 9,9-dimethyl-7,11-dioxaspiro 5.5 undecan-3-one, 1,5-dioxaspiro 5.5 undecan-9-one, 3,3-dimethyl, pubchem11226, maybridge1_004236, 1,4-cyclohexanedionemono-2,2-dimethyltrimethyleneketal
PubChem CID 587968
IUPAC Name 3,3-dimethyl-1,5-dioxaspiro[5.5]undecan-9-one
SMILES CC1(COC2(CCC(=O)CC2)OC1)C

Specifications

Melting Point 51°C
Color White
Quantity 25 g
Formula Weight 198.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Cyclohexanedione Mono-2,2-dimethyltrimethylene Ketal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.