Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Cyclohexanedione 98.0+%, TCI America™
SDP

Catalog No. C0730500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0730500G 500 g
C073025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0730500G Supplier TCI America Supplier No. C0730500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 637-88-7
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.128
MDL Number MFCD00001606
InChI Key DCZFGQYXRKMVFG-UHFFFAOYSA-N
Synonym 1,4-cyclohexanedione, tetrahydroquinone, 1,4-dioxocyclohexane, 1,4-cyclohexandione, 1,4 cyclohexane dione, 1,4-cyclohexane-dione, cyclohexan-1,4-dione, pubchem13686, cyclohexane-1,4-quinone, acmc-1b8ar
PubChem CID 12511
ChEBI CHEBI:28286
IUPAC Name cyclohexane-1,4-dione
SMILES C1CC(=O)CCC1=O

Specifications

Melting Point 78°C
Color White-Yellow
Boiling Point 132°C
Quantity 500 g
Formula Weight 112.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Cyclohexanedione
EINECSNumber (3)-3893
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.