Learn More
Medchemexpress LLC 1,4-d-gulonolactone | 1615286 | MFCD00005391 | 99.6% | 178.14 g/mol | C6H10O6 | 25 G
SDP
Catalog No. 5000307906
Encompass_Preferred
1,4-d-gulonolactone is an endogenous metabolite provided as a high-purity analytical and research standard for biochemical and analytical applications. It is supplied as a stable solid suitable for use in method development, analytical validation, and biochemical studies.
  • Endogenous metabolite and precursor in ascorbate biosynthesis.
  • High purity (99.56%) suitable for analytical use.
  • Molecular formula C6H10O6 and molecular weight 178.14 g/mol.
  • Available in multiple pack sizes, including 25 g, for scalability.
  • Supplied as a stable solid for convenient storage and handling.
Catalog No. 50-003-07906 Supplier Medchemexpress LLC Supplier No. HY11884025G
Add to Cart
Add to Cart
1,4-d-gulonolactone is an endogenous metabolite provided as a high-purity analytical and research standard for biochemical and analytical applications. It is supplied as a stable solid suitable for use in method development, analytical validation, and biochemical studies.
  • Endogenous metabolite and precursor in ascorbate biosynthesis.
  • High purity (99.56%) suitable for analytical use.
  • Molecular formula C6H10O6 and molecular weight 178.14 g/mol.
  • Available in multiple pack sizes, including 25 g, for scalability.
  • Supplied as a stable solid for convenient storage and handling.