Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Di[[2,2':6',2″-terpyridin]-4'-yl]benzene 97.0+%, TCI America™
SDP

Catalog No. B4194200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4194200MG 200 mg
B41941G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4194200MG Supplier TCI America Supplier No. B4194200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 146406-75-9
Molecular Formula C36H24N6
Molecular Weight (g/mol) 540.63
MDL Number MFCD02093699
InChI Key NXMHYAQTBVTLRK-UHFFFAOYSA-N
PubChem CID 5103711
IUPAC Name 6-(pyridin-2-yl)-4-{4-[6-(pyridin-2-yl)-[2,2'-bipyridin]-4-yl]phenyl}-2,2'-bipyridine
SMILES C1=CC=C(N=C1)C1=CC(=CC(=N1)C1=CC=CC=N1)C1=CC=C(C=C1)C1=CC(=NC(=C1)C1=CC=CC=N1)C1=CC=CC=N1

Specifications

Melting Point 250°C
Quantity 200 mg
Formula Weight 540.63
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Di[[2,2′:6′,2′′-terpyridin]-4′-yl]benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.