Learn More
Medchemexpress LLC (-)-1,4-di-O-tosyl-2,3-O-isopropylidene-L-threitol | 37002-45-2 | MFCD00003212 | 470.56 g·mol⁻¹ | C21H26O8S2 | 1 G
SDP
Catalog No. 5000301902
Encompass_Preferred
(-)-1,4-Di-O-tosyl-2,3-O-isopropylidene-L-threitol is a chemically protected derivative of L-threitol used as a synthetic intermediate and reagent in glycobiology and carbohydrate chemistry. The tosyl and isopropylidene protections enable selective functionalization in multi-step syntheses. Consult the product data sheet and SDS for batch-specific purity and safety and handling information.
  • Protected di-tosyl derivative of L-threitol suitable for synthetic transformations
  • Useful intermediate in carbohydrate synthesis and glycobiology research
  • Molecular formula C21H26O8S2; molecular weight 470.56 g·mol⁻¹
  • Provided in laboratory pack sizes for bench-scale synthesis
  • Refer to the data sheet and SDS for purity, analytical data, and safety handling
Catalog No. 50-003-01902 Supplier Medchemexpress LLC Supplier No. HYW0390171G
(-)-1,4-Di-O-tosyl-2,3-O-isopropylidene-L-threitol is a chemically protected derivative of L-threitol used as a synthetic intermediate and reagent in glycobiology and carbohydrate chemistry. The tosyl and isopropylidene protections enable selective functionalization in multi-step syntheses. Consult the product data sheet and SDS for batch-specific purity and safety and handling information.
  • Protected di-tosyl derivative of L-threitol suitable for synthetic transformations
  • Useful intermediate in carbohydrate synthesis and glycobiology research
  • Molecular formula C21H26O8S2; molecular weight 470.56 g·mol⁻¹
  • Provided in laboratory pack sizes for bench-scale synthesis
  • Refer to the data sheet and SDS for purity, analytical data, and safety handling