Learn More
Medchemexpress LLC (-)-1,4-di-O-tosyl-2,3-O-isopropylidene-L-threitol | 37002-45-2 | MFCD00003212 | >97.0% | 470.56 | C21H26O8S2 | 500 MG
SDP
Catalog No. 5000301207
Encompass_Preferred
(-)-1,4-Di-O-tosyl-2,3-O-isopropylidene-L-threitol is a tosylated, isopropylidene-protected derivative of L-threitol used as an intermediate in carbohydrate synthesis and glycochemistry research. It functions as a protected diol for selective transformations and is suitable for synthetic routes involving protecting-group manipulations and derivatization of sugars.
  • Tosylated and isopropylidene-protected structure enables selective reactions.
  • Useful as an intermediate in carbohydrate and glycochemistry syntheses.
  • Suitable for protecting-group manipulations and subsequent functionalization.
  • Offered in multiple laboratory-scale sizes for synthetic flexibility.
  • Documentation and product information available from the supplier.
Catalog No. 50-003-01207 Supplier Medchemexpress LLC Supplier No. HYW039017500MG
(-)-1,4-Di-O-tosyl-2,3-O-isopropylidene-L-threitol is a tosylated, isopropylidene-protected derivative of L-threitol used as an intermediate in carbohydrate synthesis and glycochemistry research. It functions as a protected diol for selective transformations and is suitable for synthetic routes involving protecting-group manipulations and derivatization of sugars.
  • Tosylated and isopropylidene-protected structure enables selective reactions.
  • Useful as an intermediate in carbohydrate and glycochemistry syntheses.
  • Suitable for protecting-group manipulations and subsequent functionalization.
  • Offered in multiple laboratory-scale sizes for synthetic flexibility.
  • Documentation and product information available from the supplier.