Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (-)-1,4-di-O-tosyl-2,3-O-isopropylidene-L-threitol | 37002-45-2 | MFCD00003212 | >98.0% | 470.55 | C21H26O8S2 | 5 G
SDP

Catalog No. 5000299808
Encompass_Preferred

(-)-1,4-Di-O-tosyl-2,3-O-isopropylidene-L-threitol is a tosyl-protected isopropylidene derivative of L-threitol used as a synthetic intermediate in organic synthesis. It contains two p-toluenesulfonyl ester groups and an isopropylidene acetal that facilitate selective functionalization, substitution, and protecting-group strategies in multistep preparations.

  • Provides tosyl protection for primary hydroxyls to enable substitution reactions.
  • Contains an isopropylidene acetal that protects vicinal diol functionality.
  • Configured with defined stereochemistry for stereospecific transformations.
  • High purity suitable for research applications (HPLC >98%).
  • Molecular formula C21H26O8S2; molecular weight 470.55.

Catalog No. 50-002-99808 Supplier Medchemexpress LLC Supplier No. HYW0390175G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(-)-1,4-Di-O-tosyl-2,3-O-isopropylidene-L-threitol is a tosyl-protected isopropylidene derivative of L-threitol used as a synthetic intermediate in organic synthesis. It contains two p-toluenesulfonyl ester groups and an isopropylidene acetal that facilitate selective functionalization, substitution, and protecting-group strategies in multistep preparations.

  • Provides tosyl protection for primary hydroxyls to enable substitution reactions.
  • Contains an isopropylidene acetal that protects vicinal diol functionality.
  • Configured with defined stereochemistry for stereospecific transformations.
  • High purity suitable for research applications (HPLC >98%).
  • Molecular formula C21H26O8S2; molecular weight 470.55.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.