Learn More
Medchemexpress LLC (-)-1,4-di-O-tosyl-2,3-O-isopropylidene-L-threitol | 37002-45-2 | MFCD00003212 | >98.0% | 470.56 | C21H26O8S2 | 25 G
SDP
Catalog No. 5000304024
Encompass_Preferred
(-)-1,4-di-O-tosyl-2,3-O-isopropylidene-L-threitol is a chiral, tosylated, protected threitol used as a building block and intermediate in stereoselective organic synthesis and glycochemistry. It is supplied as a white to off-white solid (C21H26O8S2, MW 470.56) and is intended for research use with recommended storage at 4°C, protected from light and under inert atmosphere.
  • Chiral protected intermediate for stereoselective organic synthesis.
  • Tosyl groups enable selective activation and substitution of hydroxyls.
  • High purity (reported >98% by HPLC) suitable for research applications.
  • White to off-white solid that is easy to weigh and handle.
  • Stable with recommended refrigerated, light-protected storage under inert gas.
  • Available in multiple lab-scale pack sizes to support synthesis workflows.
Catalog No. 50-003-04024 Supplier Medchemexpress LLC Supplier No. 25GHYW039017
(-)-1,4-di-O-tosyl-2,3-O-isopropylidene-L-threitol is a chiral, tosylated, protected threitol used as a building block and intermediate in stereoselective organic synthesis and glycochemistry. It is supplied as a white to off-white solid (C21H26O8S2, MW 470.56) and is intended for research use with recommended storage at 4°C, protected from light and under inert atmosphere.
  • Chiral protected intermediate for stereoselective organic synthesis.
  • Tosyl groups enable selective activation and substitution of hydroxyls.
  • High purity (reported >98% by HPLC) suitable for research applications.
  • White to off-white solid that is easy to weigh and handle.
  • Stable with recommended refrigerated, light-protected storage under inert gas.
  • Available in multiple lab-scale pack sizes to support synthesis workflows.