Learn More
Medchemexpress LLC (-)-1,4-di-O-tosyl-2,3-O-isopropylidene-L-threitol | 37002-45-2 | MFCD00003212 | >98.0% | 470.56 g/mol | C21H26O8S2 | 10 G
SDP
Catalog No. 5000301208
Encompass_Preferred
(-)-1,4-Di-O-tosyl-2,3-O-isopropylidene-L-threitol is a tosylated, protected threitol derivative used as an intermediate and reagent in glycochemistry and carbohydrate synthesis. It is supplied as a solid for laboratory research and synthetic applications.
  • Tosylated protected threitol enabling selective substitution of hydroxyl groups.
  • Useful intermediate for glycochemistry and carbohydrate modification.
  • Available in multiple package sizes for bench-scale synthesis.
  • Reported purity >98.0% by HPLC in supplier documentation.
  • Provided as a crystalline solid suitable for storage and handling in standard laboratory conditions.
Catalog No. 50-003-01208 Supplier Medchemexpress LLC Supplier No. HYW03901710G
(-)-1,4-Di-O-tosyl-2,3-O-isopropylidene-L-threitol is a tosylated, protected threitol derivative used as an intermediate and reagent in glycochemistry and carbohydrate synthesis. It is supplied as a solid for laboratory research and synthetic applications.
  • Tosylated protected threitol enabling selective substitution of hydroxyl groups.
  • Useful intermediate for glycochemistry and carbohydrate modification.
  • Available in multiple package sizes for bench-scale synthesis.
  • Reported purity >98.0% by HPLC in supplier documentation.
  • Provided as a crystalline solid suitable for storage and handling in standard laboratory conditions.