Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (-)-1,4-di-O-tosyl-2,3-O-isopropylidene-L-threitol | 37002-45-2 | MFCD00003212, MFCD00063235 | 470.56 g·mol⁻¹ | C21H26O8S2 | 250 MG
SDP

Catalog No. 5000303570
Encompass_Preferred

(-)-1,4-Di-O-tosyl-2,3-O-isopropylidene-L-threitol is a tosylated threitol derivative used as a protected intermediate in glycochemistry and carbohydrate synthesis. It is employed as a synthetic building block for preparing substituted sugars and glycomimetics and requires protection from light and moisture during storage.

  • Used as a protected intermediate in glycochemistry and carbohydrate synthesis.
  • Applicable for preparation of substituted sugars and glycomimetics.
  • Molecular formula C21H26O8S2 and molecular weight 470.56 g·mol⁻¹.
  • Solid, white to off-white physical form.
  • Store protected from light and under inert atmosphere; in solvent: -80°C (6 months), -20°C (1 month).

Catalog No. 50-003-03570 Supplier Medchemexpress LLC Supplier No. 250MGHYW039017
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(-)-1,4-Di-O-tosyl-2,3-O-isopropylidene-L-threitol is a tosylated threitol derivative used as a protected intermediate in glycochemistry and carbohydrate synthesis. It is employed as a synthetic building block for preparing substituted sugars and glycomimetics and requires protection from light and moisture during storage.

  • Used as a protected intermediate in glycochemistry and carbohydrate synthesis.
  • Applicable for preparation of substituted sugars and glycomimetics.
  • Molecular formula C21H26O8S2 and molecular weight 470.56 g·mol⁻¹.
  • Solid, white to off-white physical form.
  • Store protected from light and under inert atmosphere; in solvent: -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.