Learn More
Chemglass Life Sciences 1/4" Dia x 12" Long PFA Coated Thermocouple Probe, Type "J"
SDP
Catalog No. CG320057
Encompass_Preferred
1/4" Dia x 12" Long PFA Coated Thermocouple Probe, Type "J". For use with Temperature Controllers CG-15001, CG-3200, CG-3202, CG-3404, CG-3206, CG-3209, and CG-3210. Thermocouples have a standard male plug (OST) and are all PFA COATED.Maximum Probe Temperature Based on Coating- FEP is 200¿C and the Black PFA is 250¿C
Catalog No. CG320057 Supplier Chemglass Life Sciences Supplier No. CG320057
1/4" Dia x 12" Long PFA Coated Thermocouple Probe, Type "J". For use with Temperature Controllers CG-15001, CG-3200, CG-3202, CG-3404, CG-3206, CG-3209, and CG-3210. Thermocouples have a standard male plug (OST) and are all PFA COATED.Maximum Probe Temperature Based on Coating- FEP is 200¿C and the Black PFA is 250¿C