Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Diamino-2,3-dicyano-9,10-anthraquinone 90.0+%, TCI America™
SDP

Catalog No. D214825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D214825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D214825G Supplier TCI America Supplier No. D214825G

Chemical Identifiers

CAS 81-41-4
Molecular Formula C16H8N4O2
Molecular Weight (g/mol) 288.27
MDL Number MFCD00053082
InChI Key QUZJFTXRXJQLBH-UHFFFAOYSA-N
Synonym 1,4-Diamino-9,10-anthraquinone-2,3-dicarbonitrile
PubChem CID 66477
IUPAC Name 1,4-diamino-9,10-dioxo-9,10-dihydroanthracene-2,3-dicarbonitrile
SMILES NC1=C(C#N)C(C#N)=C(N)C2=C1C(=O)C1=CC=CC=C1C2=O

Specifications

Color Blue-Purple
Quantity 25 g
Formula Weight 288.27
Percent Purity ≥90.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Diamino-2,3-dicyano-9,10-anthraquinone
EINECSNumber (4)-0713
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.